Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177390.3 | NP_796364.2 | 4 - 5 | 802 | 79 | |
GenBank mRNA | AK037051.1 | - | 4 - 5 | 781 | 79 | |
AK049374.1 | - | 4 - 5 | 803 | 79 | ||
BC150765.1 | - | 4 - 5 | 668 | 9087676 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |