Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178381.3 | NP_848468.2 | 22 - 23 | 2192 | 69 | |
GenBank mRNA | AK017334.1 | - | 3 - 4 | 242 | 69 | |
BC025477.1 | - | 21 - 22 | 2268 | 69 | ||
BC049815.1 | - | 11 - 12 | 2298 | 69 | ||
BY717147.1 | - | 3 - 4 | 238 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |