Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178386.3 | NP_848473.2 | 3 - 4 | 608 | 139 | |
GenBank mRNA | AK006599.1 | - | 3 - 4 | 609 | 139 | |
AK132722.1 | - | 3 - 4 | 609 | 139 | ||
AY550241.1 | - | 3 - 4 | 547 | 139 | ||
BC050810.1 | - | 3 - 4 | 547 | 6772660 | 139 | |
BC139243.1 | - | 3 - 4 | 543 | 8862265 | 139 | |
BC139245.1 | - | 3 - 4 | 543 | 8862267 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |