Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gale | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178389.3 | NP_848476.1 | 8 - 9 | 1049 | 92 | |
GenBank mRNA | AK009083.1 | - | 9 - 10 | 917 | 92 | |
AK146184.1 | - | 9 - 10 | 940 | 92 | ||
BC027438.1 | - | 8 - 9 | 1049 | 4224021 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |