Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178608.4 | NP_848723.1 | 3 - 4 | 469 | 78 | |
GenBank mRNA | AK035238.1 | - | 3 - 4 | 303 | 78 | |
AK045056.1 | - | 3 - 4 | 291 | 78 | ||
AK053158.1 | - | 3 - 4 | 287 | 78 | ||
AK136916.1 | - | 3 - 4 | 248 | 78 | ||
AY562229.1 | - | 3 - 4 | 180 | 78 | ||
BC046826.1 | - | 3 - 4 | 240 | 6417744 | 78 | |
CX239705.1 | - | 3 - 4 | 469 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |