Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27452131 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27452135 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27452136 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27452134 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27452130 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27452132 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P174736 |
AK078627.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P174736 |
AK032472.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P174736 |
AK036447.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P174736 |
NM_178610.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1957319 |
BC094236.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1957319 |
AK168675.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1957319 |
AK078627.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1957319 |
AK146926.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1957319 |
AK032472.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
82741 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
510606 |
AB013819.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
510606 |
AF077349.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
510606 |
AK142768.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
510606 |
BC004702.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
510606 |
EU366955.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
510606 |
NM_001012273.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
510606 |
NM_009689.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
734752 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
753490 |
AK053959.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
146504 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
878731 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
319970 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
387706 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
86900 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
83557 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
174697 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P100830 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P177051 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1957319 |
BC094236.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1957319 |
AK168675.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1957319 |
AK078627.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1957319 |
AK146926.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1957319 |
AK032472.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P174736 |
AK078627.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P174736 |
AK032472.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P174736 |
AK036447.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P174736 |
NM_178610.4 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452120 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452119 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452137 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452129 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452147 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452139 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452124 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452140 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452143 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452136 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452127 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452111 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452130 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452126 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452115 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452141 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452131 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452132 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452113 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452110 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452134 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452122 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452116 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452150 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452149 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452118 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27452109 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07194620 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P177051 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P100830 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P169507 |
BC094236.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P169507 |
AK168675.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P169507 |
BC089510.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P169507 |
AK078627.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P169507 |
AK146926.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P86917 |
AK078627.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P86917 |
NM_178610.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P357241 |
AK045526.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P555382 |
BC094236.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P555382 |
AK168675.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P555382 |
BC089510.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P555382 |
AK041854.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P555382 |
AK078627.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P138537 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P177051 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P100830 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08863539 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08618555 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08935652 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08935651 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08618553 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08863538 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08618549 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27452130 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27452131 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27452135 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27452132 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27452134 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27452136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452230 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452274 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452194 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452264 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452192 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452224 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452176 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452105 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452286 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452164 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452281 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452132 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452244 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452255 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452273 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452206 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452143 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452275 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452208 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452187 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452160 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452258 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452169 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452197 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452188 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452171 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452238 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452259 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452155 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452185 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452225 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452232 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452272 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452175 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452200 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452213 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452173 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452162 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452120 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452096 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452289 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452287 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452109 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452236 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452276 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452170 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452307 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452243 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452284 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452219 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452134 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452306 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452229 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452226 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452205 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452295 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452254 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452290 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452211 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452149 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452311 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452299 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452215 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452163 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452129 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452233 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452202 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452250 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452297 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452309 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452270 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452159 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452166 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452239 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452237 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452267 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452198 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452107 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452141 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452223 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452097 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452265 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452283 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452241 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452252 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452278 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452147 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452190 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452203 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27452246 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452109 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452113 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452141 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452176 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08373819 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452143 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452156 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08662747 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452188 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452120 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08615614 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452178 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452169 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452170 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452175 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452094 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452105 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452130 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452139 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452136 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452171 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08590877 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08935652 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452132 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452159 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08801389 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452190 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452122 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452107 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452118 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08202758 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452162 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08590865 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452124 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452192 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452166 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452101 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452186 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452116 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08898591 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452128 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08590874 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452111 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452150 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452146 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452155 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452097 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452129 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452131 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452149 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08662750 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08304372 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452092 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452185 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452134 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452187 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452110 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452090 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452100 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08590867 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452115 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27452127 |
- |
- |