Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178614.4 | NP_848729.1 | 8 - 9 | 903 | 102 | |
GenBank mRNA | AK003990.1 | - | 8 - 9 | 858 | 102 | |
AK039949.1 | - | 8 - 9 | 903 | 102 | ||
AK041783.1 | - | 897 | 102 | |||
AK084395.1 | - | 8 - 9 | 897 | 102 | ||
AK086633.1 | - | 897 | 102 | |||
AK144975.1 | - | 8 - 9 | 904 | 102 | ||
AK159740.1 | - | 8 - 9 | 915 | 102 | ||
AK161378.1 | - | 8 - 9 | 893 | 102 | ||
AK167806.1 | - | 8 - 9 | 897 | 102 | ||
AK168510.1 | - | 8 - 9 | 859 | 102 | ||
AK169066.1 | - | 8 - 9 | 893 | 102 | ||
AK169351.1 | - | 8 - 9 | 860 | 102 | ||
AK215615.1 | - | 1 - 2 | 34 | 102 | ||
BC119178.1 | - | 8 - 9 | 897 | 8733927 | 102 | |
BC119180.1 | - | 8 - 9 | 897 | 8733929 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |