Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sccpdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178653.3 | NP_848768.1 | 5 - 6 | 689 | 73 | |
GenBank mRNA | AK042919.1 | - | 5 - 6 | 577 | 73 | |
AK046033.1 | - | 5 - 6 | 690 | 73 | ||
AK050554.1 | - | 5 - 6 | 638 | 73 | ||
AK077695.1 | - | 5 - 6 | 639 | 73 | ||
AK082803.1 | - | 5 - 6 | 574 | 73 | ||
AK086448.1 | - | 5 - 6 | 712 | 73 | ||
AK086632.1 | - | 5 - 6 | 687 | 73 | ||
AK145399.1 | - | 5 - 6 | 673 | 73 | ||
AK166018.1 | - | 5 - 6 | 601 | 73 | ||
BC025803.1 | - | 5 - 6 | 686 | 5100552 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |