Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp6nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080548.1 | NP_001074017.1 | 12 - 13 | 1110 | 144 | |
NM_181399.3 | NP_852064.2 | 11 - 12 | 949 | 144 | ||
GenBank mRNA | AK084045.1 | - | 12 - 13 | 934 | 144 | |
AK129037.1 | - | 12 - 13 | 962 | 144 | ||
AK133119.1 | - | 3 - 4 | 188 | 144 | ||
AK155073.1 | - | 12 - 13 | 1111 | 144 | ||
BC051184.1 | - | 11 - 12 | 952 | 144 | ||
CN455065.1 | - | 4 - 5 | 339 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |