Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181400.3 | NP_852065.2 | 8 - 9 | 2027 | 71 | |
GenBank mRNA | AK028113.1 | - | 2 - 3 | 418 | 71 | |
BC040337.1 | - | 8 - 9 | 1888 | 3586908 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |