Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025581.4 | NP_079857.3 | 1 - 2 | 292 | 134 | |
GenBank mRNA | AK172500.1 | - | 1 - 2 | 195 | 134 | |
BC019940.1 | - | 1 - 2 | 143 | 3709649 | 134 | |
DQ926864.1 | - | 1 - 2 | 234 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |