Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176940.5 | NP_795914.3 | 6 - 7 | 2273 | 74 | |
GenBank mRNA | AK034868.1 | - | 1972 | 74 | ||
AK038792.1 | - | 5 - 6 | 1966 | 74 | ||
AK087622.1 | - | 6 - 7 | 2278 | 74 | ||
AK162888.1 | - | 6 - 7 | 2268 | 74 | ||
BC082552.1 | - | 6 - 7 | 2213 | 74 | ||
BC144786.1 | - | 4 - 5 | 1863 | 9053334 | 74 | |
BC144787.1 | - | 4 - 5 | 1863 | 74 | ||
BC145703.1 | - | 4 - 5 | 1863 | 40131006 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |