Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178912.3 | NP_849243.2 | 10 - 11 | 1841 | 113 | |
GenBank mRNA | AK050306.1 | - | 10 - 11 | 1842 | 113 | |
AK053715.1 | - | 10 - 11 | 1841 | 113 | ||
AK084697.1 | - | 10 - 11 | 1845 | 113 | ||
AK173211.1 | - | 10 - 11 | 1820 | 113 | ||
BC150785.1 | - | 10 - 11 | 1835 | 9087696 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |