Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xkr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176951.5 | NP_795925.1 | 1 - 2 | 437 | 66 | |
GenBank mRNA | AK030692.1 | - | 1 - 2 | 422 | 66 | |
AK045098.1 | - | 1 - 2 | 416 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |