Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrbk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285806.1 | NP_001272735.1 | 5 - 6 | 540 | 92 | |
NM_177078.4 | NP_796052.2 | 5 - 6 | 501 | 92 | ||
GenBank mRNA | AK048763.1 | - | 5 - 6 | 466 | 92 | |
AK076533.1 | - | 5 - 6 | 399 | 92 | ||
AK134672.1 | - | 5 - 6 | 478 | 92 | ||
AK141186.1 | - | 5 - 6 | 519 | 92 | ||
AK172513.1 | - | 5 - 6 | 453 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mus musculus Probe spans exons |