Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166685.1 | NP_001160157.1 | 3 - 4 | 363 | 145 | |
NM_177088.3 | NP_796062.2 | 3 - 4 | 363 | 145 | ||
GenBank mRNA | AK052705.1 | - | 3 - 4 | 291 | 145 | |
AK076400.1 | - | 3 - 4 | 341 | 145 | ||
BC090675.1 | - | 3 - 4 | 321 | 145 | ||
BC116308.1 | - | 3 - 4 | 329 | 40095242 | 145 | |
BC116309.1 | - | 3 - 4 | 329 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |