Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177566.3 | NP_808234.2 | 6 - 7 | 1509 | 86 | |
GenBank mRNA | AK051749.1 | - | 3 - 4 | 639 | 86 | |
AK173061.1 | - | 2 - 3 | 214 | 86 | ||
BC089365.1 | - | 6 - 7 | 1509 | 6844583 | 86 | |
BC116343.1 | - | 6 - 7 | 1485 | 40098168 | 86 | |
BC116344.1 | - | 6 - 7 | 1485 | 40098169 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |