Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177667.4 | NP_808335.1 | 4 - 5 | 960 | 81 | |
GenBank mRNA | AK028895.1 | - | 4 - 5 | 961 | 81 | |
BC096484.1 | - | 4 - 5 | 937 | 79 | ||
BC132162.1 | - | 4 - 5 | 901 | 40130439 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |