Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271879.1 | NP_001258808.1 | 5 - 6 | 504 | 114 | |
NM_177694.6 | NP_808362.2 | 6 - 7 | 546 | 114 | ||
NR_073508.1 | 6 - 7 | 539 | 114 | |||
GenBank mRNA | AB125740.1 | - | 6 - 7 | 546 | 114 | |
AB206762.1 | - | 6 - 7 | 546 | 114 | ||
AK034197.1 | - | 6 - 7 | 528 | 114 | ||
BC109163.2 | - | 5 - 6 | 426 | 40047987 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |