Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vsig8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177723.4 | NP_808391.2 | 3 - 4 | 562 | 65 | |
NR_027644.1 | 2 - 3 | 281 | 65 | |||
GenBank mRNA | AK037228.1 | - | 3 - 4 | 563 | 65 | |
BC064106.1 | - | 2 - 3 | 281 | 1528324 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |