Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177839.3 | NP_808507.2 | 9 - 10 | 2241 | 74 | |
GenBank mRNA | AF455756.1 | - | 9 - 10 | 2208 | 74 | |
AJ580920.1 | - | 8 - 9 | 2160 | 74 | ||
AK048033.1 | - | 2 - 3 | 356 | 74 | ||
BC138335.1 | - | 9 - 10 | 2202 | 8861355 | 74 | |
BC138336.1 | - | 9 - 10 | 2202 | 8861356 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |