Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dido1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291433.1 | NP_001278362.1 | 13 - 14 | 3570 | 72 | |
NM_175551.4 | NP_780760.2 | 13 - 14 | 3570 | 72 | ||
NM_177852.4 | NP_808520.2 | 13 - 14 | 3518 | 72 | ||
NR_111965.1 | 11 - 12 | 3268 | 72 | |||
GenBank mRNA | AK049707.1 | - | 7 - 8 | 2185 | 72 | |
AK129117.1 | - | 13 - 14 | 3518 | 72 | ||
AY425951.1 | - | 13 - 14 | 3472 | 72 | ||
AY425952.1 | - | 13 - 14 | 3472 | 72 | ||
BC044755.1 | - | 7 - 8 | 1669 | 72 | ||
BC138712.1 | - | 12 - 13 | 3347 | 8861734 | 72 | |
BC138713.1 | - | 12 - 13 | 3347 | 8861735 | 72 | |
BC145401.1 | - | 11 - 12 | 3188 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |