Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178070.4 | NP_835171.2 | 17 - 18 | 1600 | 77 | |
GenBank mRNA | AK032076.1 | - | 17 - 18 | 1602 | 77 | |
AK189330.1 | - | 5 - 6 | 257 | 77 | ||
BC011059.1 | - | 17 - 18 | 1592 | 77 | ||
BC028783.1 | - | 7 - 8 | 451 | 77 | ||
BC034170.1 | - | 17 - 18 | 1312 | 4971695 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |