Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122822.1 | NP_001116294.1 | 2 - 3 | 180 | 130 | |
NM_011721.4 | NP_035851.3 | 2 - 3 | 290 | 130 | ||
GenBank mRNA | AF091215.1 | - | 2 - 3 | 244 | 130 | |
AF241636.1 | - | 2 - 3 | 305 | 130 | ||
AK143778.1 | - | 2 - 3 | 268 | 130 | ||
AK153824.1 | - | 3 - 4 | 373 | 130 | ||
AK158180.1 | - | 2 - 3 | 151 | 130 | ||
AK165679.1 | - | 2 - 3 | 537 | 130 | ||
AK171490.1 | - | 2 - 3 | 291 | 130 | ||
BC050921.1 | - | 2 - 3 | 235 | 6408901 | 130 | |
BC060700.1 | - | 2 - 3 | 235 | 6408901 | 130 | |
D86526.1 | - | 2 - 3 | 260 | 130 | ||
D86527.1 | - | 2 - 3 | 260 | 130 | ||
U97045.1 | - | 198 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |