Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2s3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012010.3 | NP_036140.1 | 2 - 3 | 154 | 96 | |
GenBank mRNA | AJ006587.1 | - | 2 - 3 | 144 | 96 | |
AK128949.1 | - | 2 - 3 | 153 | 96 | ||
AK146900.1 | - | 2 - 3 | 195 | 96 | ||
AK150095.1 | - | 2 - 3 | 161 | 96 | ||
AK165866.1 | - | 2 - 3 | 156 | 96 | ||
AK171317.1 | - | 2 - 3 | 155 | 96 | ||
BC036993.1 | - | 2 - 3 | 152 | 96 | ||
BC063755.1 | - | 2 - 3 | 154 | 6513251 | 96 | |
BC139234.1 | - | 2 - 3 | 154 | 8862256 | 96 | |
BC139235.1 | - | 2 - 3 | 154 | 8862257 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |