Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025988.2 | NP_080264.1 | 7 - 8 | 1048 | 82 | |
GenBank mRNA | AK008207.1 | - | 1 - 2 | 110 | 82 | |
AK008243.1 | - | 5 - 6 | 597 | 82 | ||
AK079604.1 | - | 6 - 7 | 917 | 83 | ||
AK148438.1 | - | 543 | 82 | |||
AK165381.1 | - | 1 - 2 | 185 | 82 | ||
BC048371.1 | - | 6 - 7 | 962 | 6396076 | 82 | |
BI734653.1 | - | 6 - 7 | 582 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |