Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109626.1 | NP_001103096.1 | 10 - 11 | 3015 | 75 | |
NM_001109628.1 | NP_001103098.1 | 10 - 11 | 3015 | 75 | ||
NM_026952.2 | NP_081228.2 | 10 - 11 | 3015 | 75 | ||
GenBank mRNA | AK049376.1 | - | 10 - 11 | 75 | ||
AK129237.1 | - | 10 - 11 | 2154 | 75 | ||
AK137734.1 | - | 8 - 9 | 885 | 75 | ||
AY072295.1 | - | 10 - 11 | 3006 | 75 | ||
BC116645.1 | - | 10 - 11 | 2998 | 40086381 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |