Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027256.2 | NP_081532.1 | 3 - 4 | 515 | 70 | |
GenBank mRNA | AK088635.1 | - | 3 - 4 | 459 | 70 | |
AK163383.1 | - | 3 - 4 | 469 | 70 | ||
BC013710.1 | - | 3 - 4 | 461 | 3156446 | 70 | |
CA450656.1 | - | 3 - 4 | 385 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |