Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027427.2 | NP_081703.1 | 4 - 5 | 277 | 90 | |
GenBank mRNA | AK011843.1 | - | 4 - 5 | 299 | 90 | |
AK051568.1 | - | 4 - 5 | 278 | 90 | ||
AK078205.1 | - | 4 - 5 | 289 | 90 | ||
AK146331.1 | - | 4 - 5 | 240 | 90 | ||
AK166550.1 | - | 4 - 5 | 270 | 90 | ||
BC137591.1 | - | 4 - 5 | 251 | 8860610 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |