Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK088814.1 | - | 5 - 6 | 624 | 74 | |
AK142658.1 | - | 5 - 6 | 624 | 74 | ||
AK145291.1 | - | 5 - 6 | 619 | 74 | ||
AK146207.1 | - | 5 - 6 | 623 | 74 | ||
AK150551.1 | - | 5 - 6 | 630 | 74 | ||
AK150777.1 | - | 5 - 6 | 588 | 74 | ||
AK151240.1 | - | 5 - 6 | 587 | 74 | ||
AK151409.1 | - | 5 - 6 | 630 | 74 | ||
AK151664.1 | - | 5 - 6 | 630 | 74 | ||
AK152338.1 | - | 5 - 6 | 630 | 74 | ||
AK152343.1 | - | 5 - 6 | 589 | 74 | ||
AK152991.1 | - | 5 - 6 | 589 | 74 | ||
AK153379.1 | - | 5 - 6 | 630 | 74 | ||
AK155905.1 | - | 5 - 6 | 624 | 74 | ||
AK159203.1 | - | 5 - 6 | 624 | 74 | ||
AK159350.1 | - | 5 - 6 | 624 | 74 | ||
AK159878.1 | - | 5 - 6 | 633 | 74 | ||
AK164347.1 | - | 5 - 6 | 536 | 74 | ||
AK168708.1 | - | 5 - 6 | 566 | 74 | ||
AY029764.1 | - | 5 - 6 | 516 | 74 | ||
BC012401.1 | - | 5 - 6 | 613 | 3967323 | 74 | |
CT010238.1 | - | 5 - 6 | 481 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |