Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081028.1 | NP_001074497.1 | 17 - 18 | 5028 | 67 | |
GenBank mRNA | AK044193.1 | - | 4 - 5 | 855 | 67 | |
AK087819.1 | - | 4 - 5 | 1013 | 67 | ||
AK144537.1 | - | 2 - 3 | 273 | 67 | ||
AK172968.1 | - | 10 - 11 | 1771 | 67 | ||
BC095969.1 | - | 7 - 8 | 1561 | 67 | ||
BC116408.1 | - | 2 - 3 | 621 | 67 | ||
BC128493.1 | - | 2 - 3 | 621 | 67 | ||
BC144792.1 | - | 4 - 5 | 934 | 67 | ||
BC145739.1 | - | 4 - 5 | 934 | 67 | ||
BC152360.1 | - | 2 - 3 | 621 | 67 | ||
BC152361.1 | - | 2 - 3 | 609 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |