Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174852.3 | NP_777277.2 | 10 - 11 | 2708 | 84 | |
GenBank mRNA | AK050751.1 | - | 4 - 5 | 1125 | 84 | |
AK129383.1 | - | 7 - 8 | 2003 | 84 | ||
AK141796.1 | - | 11 - 12 | 2667 | 84 | ||
AK155354.1 | - | 10 - 11 | 2749 | 84 | ||
AK170155.1 | - | 10 - 11 | 2796 | 84 | ||
AK170187.1 | - | 9 - 10 | 2003 | 84 | ||
BC038275.1 | - | 2 - 3 | 576 | 84 | ||
BC043080.1 | - | 10 - 11 | 2311 | 6400617 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |