Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174852.3 | NP_777277.2 | 7 - 8 | 1627 | 73 | |
GenBank mRNA | AK050751.1 | - | 1 - 2 | 43 | 73 | |
AK129383.1 | - | 4 - 5 | 774 | 73 | ||
AK141796.1 | - | 7 - 8 | 1531 | 73 | ||
AK155354.1 | - | 7 - 8 | 1668 | 73 | ||
AK170155.1 | - | 7 - 8 | 1715 | 73 | ||
AK170187.1 | - | 6 - 7 | 922 | 73 | ||
BC043080.1 | - | 7 - 8 | 1230 | 6400617 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |