Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22695197 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22695196 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22695195 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P510939 |
AK169018.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P510939 |
NM_181391.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P510939 |
AK007844.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P510939 |
AK013200.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1963201 |
AK168647.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1963201 |
AK169018.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1963201 |
BC030366.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1963201 |
NM_001190322.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1963201 |
NM_001190323.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
562793 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
9877 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
163030 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
387754 |
NM_007511.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
387754 |
U38477.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
691677 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
581252 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
965471 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
440617 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
498362 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
226761 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
729703 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
16447 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
56557 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
42743 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
24016 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
138545 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P155275 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P120398 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P144109 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1963201 |
AK168647.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1963201 |
AK169018.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1963201 |
BC030366.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1963201 |
NM_001190322.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1963201 |
NM_001190323.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P510939 |
AK169018.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P510939 |
NM_181391.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P510939 |
AK007844.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P510939 |
AK013200.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695218 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695198 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695217 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695204 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695227 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695209 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695219 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695213 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695214 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695203 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695215 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695200 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695206 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695210 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695197 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695221 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695226 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695208 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695211 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695224 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695195 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695220 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695205 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695229 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695232 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695196 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695201 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695207 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22695222 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P144109 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P120398 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P155275 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P788229 |
AK145105.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P364970 |
AK145105.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P364970 |
AK168647.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P364970 |
AK169018.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P364970 |
BC030366.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P364970 |
NM_001190322.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P136131 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P155275 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P120398 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P144109 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03833478 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22695196 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22695197 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22695195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695211 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695224 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695288 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695253 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695265 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695280 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695286 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695267 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695275 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695252 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695277 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695227 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695232 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695238 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695276 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695247 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695233 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695250 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695235 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695198 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695218 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695260 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695197 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695241 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695279 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695282 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695220 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695201 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695229 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695203 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695217 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695200 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695255 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695254 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695270 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695204 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695268 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695243 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695206 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695245 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695272 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695273 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695226 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695213 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695215 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695259 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695248 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695222 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695191 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695257 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695237 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695285 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695214 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22695221 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04052735 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03833467 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03606312 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04086456 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695293 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695197 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695310 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03262016 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04086457 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695345 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695346 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695285 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695232 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695214 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695308 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03833468 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695224 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695241 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695211 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695222 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695233 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695339 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695240 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695307 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03758581 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695336 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695288 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695291 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695237 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695348 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695294 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03758560 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695296 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03833487 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695239 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695303 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695207 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695341 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695215 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695227 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695282 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695329 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03606315 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695213 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03606327 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695246 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695309 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695226 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695280 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695218 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695298 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695332 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03833481 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695276 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695328 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695306 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695338 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03262013 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695286 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695192 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695209 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695221 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695238 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695198 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695279 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695248 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695201 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695229 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22695205 |
- |
- |