Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FJ715771.1 | - | 4 - 5 | 635 | 96 | |
FJ715772.1 | - | 4 - 5 | 635 | 96 | ||
FJ715773.1 | - | 4 - 5 | 635 | 96 | ||
FJ715776.1 | - | 4 - 5 | 635 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |