Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025662.5 | NP_079938.1 | 5 - 6 | 525 | 109 | |
GenBank mRNA | AK037552.1 | - | 1 - 2 | 107 | 109 | |
AK043033.1 | - | 514 | 109 | |||
AK077893.1 | - | 5 - 6 | 526 | 109 | ||
AK082806.1 | - | 5 - 6 | 518 | 109 | ||
AK168666.1 | - | 507 | 109 | |||
BC023256.1 | - | 5 - 6 | 507 | 109 | ||
BC060175.1 | - | 5 - 6 | 511 | 6827521 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |