Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epsti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029495.2 | NP_083771.1 | 10 - 11 | 1086 | 59 | |
GenBank mRNA | AK077223.1 | - | 10 - 11 | 892 | 59 | |
AK077376.1 | - | 10 - 11 | 921 | 59 | ||
AK135710.1 | - | 10 - 11 | 1080 | 59 | ||
AK136055.1 | - | 10 - 11 | 1087 | 59 | ||
AY239224.1 | - | 10 - 11 | 992 | 59 | ||
BC020120.1 | - | 10 - 11 | 919 | 3988503 | 59 | |
BC021821.1 | - | 10 - 11 | 946 | 3581940 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |