Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cela1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033612.2 | NP_291090.2 | 5 - 6 | 509 | 90 | |
GenBank mRNA | AK007392.1 | - | 5 - 6 | 507 | 90 | |
BC011218.1 | - | 5 - 6 | 483 | 4208442 | 90 | |
CB951068.1 | - | 5 - 6 | 509 | 90 | ||
M27347.1 | - | 4 - 5 | 392 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |