Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133889.2 | NP_598650.2 | 3 - 4 | 205 | 80 | |
GenBank mRNA | AK037201.1 | - | 3 - 4 | 201 | 80 | |
AK144053.1 | - | 3 - 4 | 200 | 80 | ||
BC021480.1 | - | 2 - 3 | 185 | 80 | ||
BC049111.1 | - | 3 - 4 | 205 | 6415574 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |