Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173428.3 | NP_775604.3 | 21 - 22 | 3388 | 81 | |
GenBank mRNA | AJ491857.1 | - | 19 - 20 | 3019 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |