Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173428.3 | NP_775604.3 | 52 - 53 | 8033 | 56 | |
GenBank mRNA | AJ491857.1 | - | 49 - 50 | 7598 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |