Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173867.5 | NP_776292.1 | 6 - 7 | 924 | 104 | |
GenBank mRNA | AK076040.1 | - | 6 - 7 | 855 | 104 | |
AK076046.1 | - | 1155 | 104 | |||
AK129370.1 | - | 5 - 6 | 628 | 104 | ||
BC020385.1 | - | 4 - 5 | 457 | 104 | ||
BC034759.1 | - | 3 - 4 | 243 | 104 | ||
BC060624.1 | - | 6 - 7 | 878 | 104 | ||
BC086666.1 | - | 6 - 7 | 847 | 30634817 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |