Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175022.2 | NP_778187.2 | 11 - 12 | 5717 | 62 | |
GenBank mRNA | AK032894.1 | - | 8 - 9 | 2205 | 62 | |
AK173130.1 | - | 8 - 9 | 5156 | 62 | ||
BC025846.1 | - | 3 - 4 | 878 | 62 | ||
BC058674.1 | - | 8 - 9 | 4356 | 62 | ||
BC075628.1 | - | 12 - 13 | 4627 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |