Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177331.4 | NP_796305.3 | 2 - 3 | 386 | 120 | |
GenBank mRNA | AK030995.1 | - | 2 - 3 | 396 | 120 | |
AK031077.1 | - | 2 - 3 | 365 | 120 | ||
BC090653.1 | - | 2 - 3 | 386 | 30659763 | 120 | |
BC138157.1 | - | 2 - 3 | 273 | 8861176 | 120 | |
BC138158.1 | - | 2 - 3 | 273 | 8861177 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |