Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hapln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178255.3 | NP_839986.2 | 2 - 3 | 297 | 67 | |
GenBank mRNA | AK016123.1 | - | 2 - 3 | 298 | 67 | |
AK017865.1 | - | 2 - 3 | 276 | 67 | ||
AK136956.1 | - | 289 | 67 | |||
AY262757.1 | - | 2 - 3 | 297 | 67 | ||
AY269788.1 | - | 2 - 3 | 239 | 67 | ||
BC110474.2 | - | 2 - 3 | 269 | 40045754 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |