Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178617.4 | NP_848732.3 | 1 - 2 | 309 | 119 | |
GenBank mRNA | AK039240.1 | - | 1 - 2 | 125 | 119 | |
AK050307.1 | - | 2 - 3 | 206 | 119 | ||
AK081951.1 | - | 1 - 2 | 201 | 119 | ||
AY278200.2 | - | 1 - 2 | 292 | 119 | ||
BC067055.1 | - | 1 - 2 | 309 | 30545678 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |