Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178877.6 | NP_849208.4 | 4 - 5 | 575 | 137 | |
GenBank mRNA | AK029417.1 | - | 2 - 3 | 254 | 137 | |
AK076505.1 | - | 5 - 6 | 616 | 137 | ||
BC090977.1 | - | 4 - 5 | 528 | 30503721 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |