Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500001532.mm.1 |
NM_181588.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001532.mm.1 |
NM_181588.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001532.mm.1 |
TC1500000207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500001533.mm.1 |
NM_181588.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001533.mm.1 |
NM_181588.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001533.mm.1 |
TC1500000207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500001535.mm.1 |
NM_181588.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001535.mm.1 |
NM_181588.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001535.mm.1 |
TC1500000207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500001534.mm.1 |
NM_181588.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001534.mm.1 |
NM_181588.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001534.mm.1 |
TC1500000207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500001531.mm.1 |
NM_181588.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001531.mm.1 |
NM_181588.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001531.mm.1 |
TC1500000207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500001529.mm.1 |
NM_181588.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001529.mm.1 |
NM_181588.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001529.mm.1 |
TC1500000207.mm.1 |
MTA TC |