Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181816.2 | NP_861537.2 | 3 - 4 | 356 | 71 | |
GenBank mRNA | AK030153.1 | - | 3 - 4 | 339 | 71 | |
BC096547.1 | - | 2 - 3 | 249 | 30360184 | 71 | |
BY069891.1 | - | 3 - 4 | 357 | 71 | ||
KC211186.1 | - | 2 - 3 | 200 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |