Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181816.2 | NP_861537.2 | 8 - 9 | 1091 | 85 | |
GenBank mRNA | AK030153.1 | - | 8 - 9 | 1073 | 85 | |
AK077147.1 | - | 3 - 4 | 469 | 85 | ||
BC096547.1 | - | 7 - 8 | 984 | 30360184 | 85 | |
KC211186.1 | - | 7 - 8 | 935 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |